LABmax Airless HF Bottle Top Dispensers
Supplier: witeg Labortechnik
5.370.823 5.370.824 5.370.825
WITG5.370.823EA 737 EUR
WITG5.370.823 WITG5.370.824 WITG5.370.825
LABmax Airless HF Bottle Top Dispensers
Bottle Top Dispensers
LABmax Airless HF is a special dispenser for hydrofluoric acid (HF). The components which have direct contact with the acid are made of HF-resistant materials. LABmax Airless HF is equipped with an automatic air-purging system. All dispensers are ready to use after no more than two dosings.
  • High quality precision instrument
  • Highest accuracy and precision, with error ranges better than DIN EN ISO 8655-5 requirements
  • 100% drip-free due to a unique anti-drip system
  • 360° rotating valve block for added safety
  • One-handed operation
  • Flexibility to use different