Heating mantles with reaction vessel, WiseTherm® WHM
Supplier: witeg Labortechnik
DH.WHM12216
WITGDH.WHM12216EA 312 EUR
WITGDH.WHM12216
Heating mantles with reaction vessel, WiseTherm® WHM
Heaters Heating Mantles
Heating mantles with aluminium exterior and fabric interior. The integrated K-type thermo-sensor is basically embedded (except fabric mantles) which enables precise control of the mantles when used together with remote digital controller. External thermo-sensor is available, which enables precise control of the temperature of liquid sample in the vessel when used together with remote digital controller.
  • Maximum temperature 450 °C
  • Can be used with magnetic stirrers
  • Acid-resistant and robust standing
  • With coil type nickel chrome heating element