Ion selective electrode, digital, NitraLyt® Plus
Supplier: WTW ONLINE
107080
WTWO107080EA 4498 EUR
WTWO107080
Ion selective electrode, digital, NitraLyt® Plus
Electrodes ISE Electrodes
With the NitraLyt® Plus 700 IQ, nitrate can be determined directly in the medium. This makes measured values available in a timely manner and they can easily be used for regulation purposes.
  • Calibration free, long-term stability
  • Economical and cost efficient
Measuring range or resolution (NO₃-N): 1 - 1000 mg/l (1 mg/l); 0,1 - 100 mg/l (0,1 mg/l)