Micropump Low-flow pump head, pressure-loaded, 0.092 mL/revolution
Supplier: Avantor Fluid Handling
Offering remote C-mount choices for your low-flow applications.
- Use for applications requiring high initial differential pressure.
- Suction-shoe design ensures complete fluid transfer for increased volumetric accuracy
- Pump efficiency increases as discharge pressure increases
The compact pump head requires a drive for proper operation. Order drives separately.<br /><br />Optional <B>Service kit</B> includes: gears, suction shoe, shaft seals, and springs. Order separately.<br /><br />Contact our Application Specialists for assistance.