Saint Gobain Transfer Tubing, Tygon® A-60-F IB, 1/2" ID x 3/4" OD; 50 Ft
Supplier: Avantor Fluid Handling
Flexibility at extreme temperatures.
- Ideal for general-purpose, single-use, and extended fluid path applications
- Single-use custom lengths and configurations available to minimize downtime
- Regulatory approvals to match your unique application
Ideal for chemical transfer, pesticide/fertilizer delivery, water feed lines, food/dairy processing, and water purification lines. The tube features excellent resistance to acidic and alkali substances, ozone, UV light, and abrasive media. It is nonconductive, heat-sealable, non-aging, and nonoxidizing. In addition, the tube maintains flexibility even at extreme temperatures. <br /><br />Temperature range is -60 to 275 °F (-51 to 135 °C).<br /><br />Can be sterilized with autoclave.