316 SS Female Reducer, 1/4" x 1/8" NPT(F) 31211-10
Supplier: Avantor Fluid Handling
Maintain a permanent connection.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Constructed of PVC with a temperature range of -10 to 140°F (-23 to 60°C)
- Sterilize via ethylene oxide only
Sturdy threaded adapters are designed for permanent installation. Choose the size and material that fits your application needs.