McMillan Flow High-Accuracy Flowmeter Power Supply, 120 VAC 60 Hz to 12 VDC
Supplier: Avantor Fluid Handling
VWR Item#MFLX32704-55
Your Organization's Price:
Ideal for use with McMillan Flow High-Accuracy Flowmeters.
  • A wide range of output options for integration into any system
These power supplies are required to power the Modular Liquid Flow Rate Sensor Systems.