Cadence 316 SS fittings; female luer x 1/4-28 UNF thread 41507-34
Supplier: Avantor Fluid Handling
VWR Item#MFLX41507-34
Your Organization's Price:
Exceptional durability.
  • Easy to disassemble for cleaning
  • Ideal for harsh chemical environments
  • Autoclavable
Constructed of durable 316 stainless steel, these Luer Adapter Fittings are ideal for a wide range of applications.