Cadence Female luer lock x 1/8" NPT, 316 SS, each 41507-46
Supplier: Avantor Fluid Handling
Exceptional durability.
- Easy to disassemble for cleaning
- Ideal for harsh chemical environments
- Autoclavable
Constructed of durable 316 stainless steel, these Luer Adapter Fittings are ideal for a wide range of applications.