Cadence 316 Stainless Steel fittings, male luer lock x male luer lock 41507-90
Supplier: Avantor Fluid Handling
Exceptional durability.
- Easy to disassemble for cleaning
- Ideal for harsh chemical environments
- Autoclavable
Constructed of durable 316 stainless steel, these Luer Adapter Fittings are ideal for a wide range of applications.