Hand-operated chemical siphon/drum pump, 12 strokes/gallon
Supplier: Avantor Fluid Handling
Ideal for most acid/base or ionic salt applications.
- Portable and self-priming
- Suitable for acid and alkali transfer
These plastic hand-operated drum pumps are ideal for water or chemical siphons. All pumps are constructed with PVC barrel and hose. Water siphon model 70607-00 includes a 60” PVC outlet hose that can be cut to fit your application. Chemical siphon models 70607-10 and 70607-12 are suitable for acid and alkali transfer.
Includes PVC barrel and hose, and PVC tube