Standard Drum Wrench, Non-Sparking Brass
Supplier: Avantor Fluid Handling
VWR Item#MFLX70607-90
Your Organization's Price:
Remove or tighten plugs on most industrial drums.
  • Fiberglass-reinforced nylon wrenches are lightweight and easy to carry
  • Use the nonsparking brass alloy models for opening drums containing flammables
  • More corrosion resistant than steel versions
  • 70607-90 and 70607-91 wrenches are made of non-sparking bronze alloy
Use the round end for outside splines of bung caps. Use the long end for internal splines of bung caps.
Round end for outside splines of bung caps. Long end for internal splines of bung caps.