Micropump Low-flow pump head, pressure-loaded, 0.042 mL/revolution
Supplier: Avantor Fluid Handling
VWR Item#MFLX07002-33
Your Organization's Price:
Offering remote C-mount choices for your low-flow applications.
  • Use for applications requiring high initial differential pressure.
  • Suction-shoe design ensures complete fluid transfer for increased volumetric accuracy
  • Pump efficiency increases as discharge pressure increases
The compact pump head requires a drive for proper operation. Order drives separately.<br /><br />Optional <B>Service kit</B> includes: gears, suction shoe, shaft seals, and springs. Order separately.<br /><br />Contact our Application Specialists for assistance.