Masterflex® Fitting, 316 Stainless Steel, Straight, Female Thread Reducer, 1/4" x 3/4" NPT(F)
Supplier: Avantor Fluid Handling
VWR Item#MFLX31410-16
Your Organization's Price:
Maintain a permanent connection.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Constructed of 316 stainless steel (SS) with a temperature range of -40 to 200°F (-40 to 93°C)
  • Sterilize via autoclave, ethylene oxide, or gamma irradiation
Sturdy threaded adapters are designed for permanent installation. Choose the size and material that fits your application needs.