Masterflex® Fittings, HDPE, Elbow, Hose Barb Union, 1/16"; 10/PK
Supplier: Avantor Fluid Handling
VWR Item#MFLX41518-47
Your Organization's Price:
Quick and easy assembly without tools.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: 1/16" ID
Create 90° bends in flexible tubing—without the kinks!