PLOT columns, CP-PoraPLOT Q and CP-PoraPLOT Q-HT
Supplier: VARIAN
CP7554 CP7551 CP7553 CP7558 CP7559 CP6953 CP7557 CP7549 CP7550 CP7552 CP7555 CP7548 CP7550I5 CP7551I5 CP7554I5 CP7556 CP6954
VARICP7554EA 0 EUR
VARICP7554 VARICP7551 VARICP7553 VARICP7558 VARICP7559 VARICP6953 VARICP7557 VARICP7549 VARICP7550 VARICP7552 VARICP7555 VARICP7548 VARICP7550I5 VARICP7551I5 VARICP7554I5 VARICP7556 VARICP6954
PLOT columns, CP-PoraPLOT Q and CP-PoraPLOT Q-HT
Chromatography Columns
CP-PoraPLOT Q is recommended for column switching systems that analyze polar and apolar volatile compounds.
  • Water elutes as a sharp peak and can therefore be quantified, improving productivity
  • Repeatable retention times for long-term stability that enhances efficiency
CP-PoraPLOT Q-HT is the high temperature version of CP-PoraPLOT Q, offering the same benefits but operating up to 290 °C.