SPE columns, AOX analysis, CHROMABOND® HR-P-AOX
Supplier: MACHEREY-NAGEL
730111.AOX 730119.AOX
MANA730111.AOXEA 206 EUR
MANA730111.AOX MANA730119.AOX
SPE columns, AOX analysis, CHROMABOND® HR-P-AOX
Chromatography Columns
CHROMABOND® HR-P-AOX is a special PS/DVB phase for the SPE enrichment of AOX from water.
  • Special polystyrene-divinylbenzene copolymer
  • Very high binding capacity for AOX (adsorbable organic halides)
Application: Extraction of AOX from water containing high salt loads or organic pollutants in accordance with DIN 38409 – H22.