TLC plates, cellulose MN 400, AVICEL®
Supplier: MACHEREY-NAGEL
801113 801123 808072 808073
552-0144EA 165 EUR
552-0144 552-0146 552-0184 552-0185
TLC plates, cellulose MN 400, AVICEL®
TLC Plates
These TLC plates and sheets are prepared by hydrolysis of high purity cellulose with HCl. They are used for carboxylic acids, lower alcohols, urea and purine derivatives.
  • Alternative selectivity to silica 60
Celluolse MN 400 (AVICEL®): Microcrystalline; average degree of polymerisation 40 to 200.