Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')ytterbium(III) ≥99%
Supplier: Thermo Fisher Scientific
|
Warning
|
39581.03
39581.06
39581.03EA
89.2
EUR
39581.03
39581.06
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')ytterbium(III) ≥99%
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')ytterbium(III)
Purity: 99%
CAS No.: 15492-52-1
Molecular Formula: C33H57O6Yb
Formula Weight: 722.86
Physical Form: Crystalline
Sensitivity: Hygroscopic
MDL No.: MFCD00000034
Formula:
C₃₃H₅₇O₆Yb Melting Pt: 167…169 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000034 CAS Number: 15492-52-1 |