Potassium diphenylphosphide 0.5 mol/l in THF, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
380768-100ML 380768-800ML
SIAL380768-100MLEA 126 EUR
SIAL380768-100ML SIAL380768-800ML
Potassium diphenylphosphide 0.5 mol/l in THF, Sigma-Aldrich®
Potassium diphenylphosphide
Formula: C₁₂H₁₀KP
MDL Number: MFCD00134530
CAS Number: 15475-27-1