HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate), Sigma-Aldrich®
Supplier: Merck
Synonyms: HATU, 1-[Bis(dimethylamino)methylene]-1H-1,2,3-triazolo[4,5-b]pyridinium 3-oxide, hexafluorophosphate
11373-1G 11373-25G 11373-5G
SIAL11373-1GEA 31.8 EUR
SIAL11373-1G SIAL11373-25G SIAL11373-5G
HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate), Sigma-Aldrich®
HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate)
Formula: C₁₀H₁₅F₆N₆OP
MDL Number: MFCD00274639
CAS Number: 148893-10-1