Ethyl 4-(bromomethyl)-2-(2-pyridinyl)-1,3-thiazole-5-carboxylate
Supplier: Cayman Chemical
13007-100 13007-250 13007-500 13007-1
CAYM13007-100EA 22 EUR
CAYM13007-100 CAYM13007-250 CAYM13007-500 CAYM13007-1
Ethyl 4-(bromomethyl)-2-(2-pyridinyl)-1,3-thiazole-5-carboxylate
Ethyl 4-(bromomethyl)-2-(2-pyridinyl)-1,3-thiazole-5-carboxylate
Formula: C₁₂H₁₁BrN₂O₂S
MDL Number: MFCD11869291
CAS Number: 1138444-37-7