Diphenylphosphine oxide, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
287881-1G
287881-5G
SIAL287881-1GEA
73.7
EUR
SIAL287881-1G
SIAL287881-5G
Diphenylphosphine oxide, Sigma-Aldrich®
Diphenylphosphine oxide
Formula:
C₁₂H₁₁OP MW: 202.20 g/mol |
MDL Number:
MFCD00002079 CAS Number: 4559-70-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...