Sodium borodeuteride (98% D) for NMR spectroscopy
Supplier: Thermo Fisher Scientific
Danger
194950010 194950050
ACRO194950010EA 71.4 EUR
ACRO194950010 ACRO194950050
Sodium borodeuteride (98% D) for NMR spectroscopy
Sodium borodeuteride
Formula: D₄BNa
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00003519
CAS Number: 15681-89-7
EINECS: 239-764-7
UN: 1426
ADR: 4.3,I
Order Now
Specification Test Results
Appearance White to off-white to light grey Powder and/or chunks
D-Enrichment ≥97.5 %