Sodium borodeuteride (98% D), Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ100

205591-1G 205591-5G
SIAL205591-1GEA 101 EUR
SIAL205591-1G SIAL205591-5G
Sodium borodeuteride (98% D), Sigma-Aldrich®
Sodium borodeuteride
Formula: D₄BNa
MW: 41.857 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003519
CAS Number: 15681-89-7
EINECS: 239-764-7
UN: 1426
ADR: 4.3 I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR