Nickel(II) tetrafluoroborate hexahydrate 99%
Supplier: Thermo Fisher Scientific
Danger
358730100 358730250
ACRO358730100EA 31.8 EUR
ACRO358730100 ACRO358730250
Nickel(II) tetrafluoroborate hexahydrate 99%
Nickel (II) tetrafluoroborate hexahydrate
Formula: B₂F₈Ni·6H₂O
MW: 340.394 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150259
CAS Number: 15684-36-3
EINECS: 238-753-4
UN: 1759
ADR: 8 II
Order Now
Specification Test Results
Appearance Blue-green Powder or crystals
Titration Complexometric ≥98.5 %