2,5-Difluoro-4-nitrobenzoic acid ≥97%
Supplier: Thermo Fisher Scientific
Warning
H33757.03 H33757.06
H33757.03EA 104 EUR
H33757.03 H33757.06
2,5-Difluoro-4-nitrobenzoic acid ≥97%
2,5-Difluoro-4-nitrobenzoic acid
Formula: C₇H₃F₂NO₄
MW: 203.10 g/mol
Melting Pt: 145…147 °C
Storage Temperature: Ambient
MDL Number: MFCD03425705
CAS Number: 116465-48-6