(±)-Aspartic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2-Aminobutanedioic acid, DL-Aspartic acid
M-Clarity™ quality level = MQ200
A9006-100G A9006-1KG A9006-500G
SIALA9006-100GEA 30.6 EUR
SIALA9006-100G SIALA9006-1KG SIALA9006-500G
(±)-Aspartic acid, Sigma-Aldrich®
(±)-Aspartic acid
Formula: C₄H₇NO₄
MW: 133.10 g/mol
MDL Number: MFCD00063083
CAS Number: 617-45-8
EINECS: 210-513-3