2'-Fluoroacetanilide
Supplier: Thermo Scientific
SPB06387.1
SPB06387.10
SPB06387.50
MAYBSPB06387.1EA
0
EUR
MAYBSPB06387.1
MAYBSPB06387.10
MAYBSPB06387.50
2'-Fluoroacetanilide
2'-Fluoroacetanilide
Formula:
C₈H₈FNO Boiling Pt: 140…142 °C (14 mmHg) Melting Pt: 81 °C |
MDL Number:
MFCD00000315 CAS Number: 399-31-5 EINECS: 206-916-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...