EGTA-Na4 (Ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid tetrasodium salt), Sigma-Aldrich®
Supplier: Merck
Synonyms: 3,12-Bis(carboxymethyl)-6,9-dioxa-3,12-diazatetradecane-1,14-dioic acid, tetrasodium salt, Ethylene glycol-bis(beta-aminoethyl ether)-N,N,N',N'-tetraacetic acid tetrasodium salt, EGTA tetrasodium
M-Clarity™ quality level = MQ200
E8145-10G E8145-50G
SIALE8145-10GEA 147 EUR
SIALE8145-10G SIALE8145-50G
EGTA-Na4 (Ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid tetrasodium salt), Sigma-Aldrich®
EGTA-Na4 (Ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid tetrasodium salt)
Formula: C₁₄H₂₀N₂Na₄O₁₀
Storage Temperature: Ambient
MDL Number: MFCD00274420
CAS Number: 13368-13-3