3-Oxetanone 95%
Supplier: Thermo Fisher Scientific
Danger
H57870.03 H57870.06
H57870.03EA 240 EUR
H57870.03 H57870.06
3-Oxetanone 95%
3-Oxetanone
Formula: C₃H₄O₂
Boiling Pt: 140 °C
Density: 1.124
Flash Pt: 26 °C (79 °F)
Storage Temperature: Freezer
MDL Number: MFCD09263255
CAS Number: 6704-31-0
UN: 1993
ADR: 3,III