Ethyl-2-amino-4-phenylthiophene-3-carboxylate
Supplier: Cayman Chemical
Synonyms: Ethyl 2-Amino-4-phenylthiophene-3-carboxylate
13387-100 13387-250 13387-500 13387-1
CAYM13387-100EA 22 EUR
CAYM13387-100 CAYM13387-250 CAYM13387-500 CAYM13387-1
Ethyl-2-amino-4-phenylthiophene-3-carboxylate
Ethyl 2-amino-4-phenylthiophene-3-carboxylate
Formula: C₁₃H₁₃NO₂S
MW: 247.32 g/mol
MDL Number: MFCD00126391
CAS Number: 4815-36-5