Gel permeation chromatography columns, ResiPore™
Supplier: VARIAN
PL1113-1300 PL1113-6300 PL1513-1300 PL1513-5300
VARIPL1113-1300EA 685 EUR
VARIPL1113-1300 VARIPL1113-6300 VARIPL1513-1300 VARIPL1513-5300
Gel permeation chromatography columns, ResiPore™
Chromatography Columns
Ideal for the analysis of resins, and condensation polymers with complex molecular weight distributions that include oligomer content.
  • Efficient separation of complex molecular weight distributions
  • Reveals oligomer content to provide a true representation of the sample
  • High pore volume extracts maximum information from the analyte
By combining a low 3 µm particle size and high pore volume, high efficiency ResiPore columns offer maximum resolution of these intermediate molecular weight polymers.