Chemical resistant gloves, Showa 727 / 730
Supplier: Showa
727-07 727-08 727-09 727-10 727-11 730-07 730-08 730-09 730-10 730-11
112-3207EA 43.1 EUR
112-3207 112-3208 112-3209 112-3210 112-3221 112-3223 112-3224 112-3227 112-3228 112-3229
Chemical resistant gloves, Showa 727 / 730
Gloves
These fully nitrile coated gloves are either available with cotton flock lining or without lining. They protect hands from oils, hydrocarbons, grease and abrasion, while providing an effective long lasting grip.
  • Textured finish provides better grip
  • Anatomical shape: Designed for easy movement and continuous wear
  • Do not contain silicones: Minimal allergy risks
  • Impermeable for working in damp or greasy environments
  • Gloves with flock lining absorb perspiration
  • Thin, unlined gloves provide a 'second skin' feel
Ideal for automotive, chemical and oil-based industries as well as for maintenance and cleaning applications.
Certifications: EN 388:2016 (2001X), EN ISO 374-1 Type A (JKLOPT), EN ISO 374-5; Cat. III
Food contact approved