Sweatshirt, Essential round neck
Supplier: FRISTADS KANSAS
1010345402XL 1010345403XL 101034540M 101034540S 101034540XL
113-6830EA 0 EUR
113-6830 113-6831 113-6833 113-6834 113-6835
Sweatshirt, Essential round neck
Workwear
Army-style sweatshirt made of 80% wool and 20% polyamide.
  • Round neck
  • Woven twill reinforcement on yoke and elbows
  • Woven twill epaulets, woven twill pen pocket on left sleeve
Ideal for use in service industries.
Order Now
SPECIFICATIONS
Model Round neck
Environmentally Preferable
Garment application Services,General Purpose
Risk area Cold
Gender For men
Sleeve or leg length Long
Fabric 80% wool, 20% polyamide