3-(1-Piperidinyl)phenol
Supplier: Thermo Scientific
S12998.250
S12998.1
MAYBS12998.250EA
0
EUR
MAYBS12998.250
MAYBS12998.1
3-(1-Piperidinyl)phenol
3-(1-Piperidinyl)phenol
Formula:
C₁₁H₁₅NO |
MDL Number:
MFCD00053001 CAS Number: 27292-50-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...