Potassium iodate standardised solution 0.1 N in aqueous solution
Supplier: Thermo Fisher Scientific
35586.K2 35586.K7
35586.K2EA 62 EUR
35586.K2 35586.K7
Potassium iodate standardised solution 0.1 N in aqueous solution
Potassium iodate
CAS No.: 7758-05-6 Molecular Formula: KIO3 Formula Weight: 214.00 Physical Form: Liquid Appearance: Colourless Odour: Odourless MDL No.: MFCD00011406
Formula: KlO₃
MW: 214 g/mol
Melting Pt: 560
Storage Temperature: Ambient
MDL Number: MFCD00011406
CAS Number: 7758-05-6
EINECS: 231-831-9
Merck Index: 13,07726