N,N-Bis(trifluoromethanesulphonyl)aniline, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1,1,1-Trifluoro-N-phenyl-N-((trifluoromethyl)sulphonyl)methanesulphonamide, N-Phenylbis(trifluoromethanesulphonimide)
M-Clarity™ quality level = MQ200
295973-25G 295973-5G
SIAL295973-25GEA 190 EUR
SIAL295973-25G SIAL295973-5G
N,N-Bis(trifluoromethanesulphonyl)aniline, Sigma-Aldrich®
N,N-Bis(trifluoromethanesulphonyl)aniline
Formula: C₈H₅F₆NO₄S₂
MW: 357.25 g/mol
MDL Number: MFCD00000404
CAS Number: 37595-74-7