Sodium-1-hexadecanesulphonate ≥97%
Supplier: Thermo Fisher Scientific
Warning
B20382.06 B20382.14 B20382.22
B20382.06EA 45.7 EUR
B20382.06 B20382.14 B20382.22
Sodium-1-hexadecanesulphonate ≥97%
Sodium 1-hexadecanesulphonate
Purity: 97% CAS No.: 15015-81-3 Molecular Formula: C16H33SO3Na Formula Weight: 328.49 Beilstein Reference: 3920654 MDL No.: MFCD00007528
Formula: C₁₆H₃₃NaSO₃
Storage Temperature: Ambient
MDL Number: MFCD00007528
CAS Number: 15015-81-3
EINECS: 239-100-6