4-Bromo-2-ethylaniline hydrochloride

Supplier: Thermo Scientific

TL00569.1 TL00569.10 TL00569.50
MAYBTL00569.1EA 0 EUR
MAYBTL00569.1 MAYBTL00569.10 MAYBTL00569.50
4-Bromo-2-ethylaniline hydrochloride
4-Bromo-2-ethylaniline hydrochloride
Formula: C₈H₁₁BrClN
MDL Number: MFCD00156127
CAS Number: 30273-22-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR