4'-Hydroxynonanophenone 96%
Supplier: Thermo Fisher Scientific
|
Warning
|
L02293.06
L02293.14
L02293.06EA
64.8
EUR
L02293.06
L02293.14
4'-Hydroxynonanophenone 96%
4'-Hydroxynonanophenone
Purity: 96%
CAS No.: 14392-69-9
Molecular Formula: C15H22O2
Formula Weight: 234.34
Melting Point: 53-57°
Beilstein Reference: 2103401
Formula:
C₁₅H₂₂O₂ Melting Pt: 53…57 °C Storage Temperature: Ambient |
MDL Number:
MFCD00020174 CAS Number: 14392-69-9 |