Guanidinium hydrochloride 99%, Ultrapure
Supplier: Thermo Fisher Scientific
|
Warning
|
Purity: 99%
CAS No.: 50-01-1
Molecular Formula: CH5N3.HCl
Formula Weight: 95.53
Physical Form: Powder
Density: 1.344
Appearance: White
Melting Point: 185-188°
Beilstein Reference: 3591990
Merck Reference: 14,4562
MDL No.: MFCD00013026
Formula:
NH₂C(=NH)NH₂·HCl Melting Pt: 185…188 °C Density: 1.344 Storage Temperature: Ambient |
MDL Number:
MFCD00013026 CAS Number: 50-01-1 EINECS: 200-002-3 Merck Index: 14,4562 |