L(+)-Asparagine monohydrate cell culture reagent
Supplier: Thermo Fisher Scientific
Synonyms: L-Asparagine monohydrate, H-Asn-OH·H₂O, (S)-(+)-Asparagine monohydrate, (S)-2,4-Diamino-4-oxobutanoic acid hydrate, H-Asn-OH.H2O
J62869.22 J62869.A1 J62869.36
J62869.22EA 88 EUR
J62869.22 J62869.A1 J62869.36
L(+)-Asparagine monohydrate cell culture reagent
L(+)-Asparagine monohydrate
CAS No.: 5794-13-8 Molecular Formula: C4H8N2O3.H2O Formula Weight: 150.14 (132.12anhy) Physical Form: Powder Appearance: White to off-white Melting Point: 215-217° Beilstein Reference: 1723527 MDL No.: MFCD00151038
Formula: C₄H₈N₂O₃·1H₂O
MW: 150.13 g/mol
Melting Pt: 215…217 °C
Density: 1.567 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00151038
CAS Number: 5794-13-8
EINECS: 200-735-9
Merck Index: 14,00837