Sodium hydroxide 10 mol/l (10 N) in aqueous solution
Supplier: Thermo Fisher Scientific
Danger
J63736.AE J63736.AK
J63736.AEEA 50.7 EUR
J63736.AE J63736.AK
Sodium hydroxide 10 mol/l (10 N) in aqueous solution
Sodium hydroxide
Formula: NaOH
Boiling Pt: 109 °C (1013 hPa)
Density: ≤1.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II