Calcium chloride 100 mM in aqueous solution, sterile-filtered
Supplier: Thermo Fisher Scientific
J62905.AP
J62905.K2
J62905.APEA
94.5
EUR
J62905.AP
J62905.K2
Calcium chloride 100 mM in aqueous solution, sterile-filtered
Calcium chloride
CAS No.: 10043-52-4
Molecular Formula: CaCl2
Formula Weight: 110.99
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Formula:
CaCl₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD00010903 CAS Number: 10043-52-4 EINECS: 233-140-8 |