1-Methyl-3-octylimidazolium bis(trifluoromethylsulphonyl)imide ≥99%
Supplier: Thermo Fisher Scientific
Synonyms: 1-Methyl-3-n-octylimidazolium Bis(trifluoromethanesulphonyl)imide
H35352.06 H35352.14 H35352.22
H35352.06EA 63.9 EUR
H35352.06 H35352.14 H35352.22
1-Methyl-3-octylimidazolium bis(trifluoromethylsulphonyl)imide ≥99%
1-Methyl-3-octylimidazolium bis(trifluoromethylsulphonyl)imide
Formula: C₁₄H₂₃F₆N₃O₄S₂
MW: 475.48 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08458946
CAS Number: 178631-04-4