Lidocaine hydrochloride monohydrate ≥98%
Supplier: Thermo Fisher Scientific
Danger
J63035.06 J63035.14
J63035.06EA 40.6 EUR
J63035.06 J63035.14
Lidocaine hydrochloride monohydrate ≥98%
Lidocaine hydrochloride monohydrate
Formula: C₁₄H₂₃ClN₂O·1H₂O
Melting Pt: 75…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00150329
CAS Number: 6108-05-0
UN: 2811
ADR: 6.1,III
Merck Index: 14,5482