1-Aminobenzotriazole 97%
Supplier: Thermo Fisher Scientific
|
Warning
|
Purity: 98%
CAS No.: 1614-12-6
Molecular Formula: C6H6N4
Formula Weight: 134.14
Physical Form: Powder
Appearance: White to pale brown
Melting Point: 81-85°
Solubility: Soluble in chloroform, DMSO, ethanol or water
Beilstein Reference: 607843
MDL No.: MFCD00132902
Suicide inhibitor of cytochrome P450 and chloroperoxidase
Formula:
C₆H₆N₄ Melting Pt: 81…85 °C Storage Temperature: Ambient |
MDL Number:
MFCD00132902 CAS Number: 1614-12-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...