3'-Deoxyadenosine (cordycepine) ≥98%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: 3'-Deoxyadenosine, (2R,3R,5S)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3-ol, Cordycepin
J65366.MB J65366.MC
J65366.MBEA 174 EUR
J65366.MB J65366.MC
3'-Deoxyadenosine (cordycepine) ≥98%
3'-Deoxyadenosine (Cordycepine)
Formula: C₁₀H₁₃N₅O₃
MW: 251.24 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00037998
CAS Number: 73-03-0
EINECS: 200-791-4
UN: 2811
ADR: 6.1 III
Merck Index: 14,2529