Sample containers, straight, with screw cap, WHEATON®
Supplier: DWK Life Sciences
360200 360205 360207 360251 360252 360199 360201 360206 360250
216-1013EA 189 EUR
216-1013 216-1015 216-1017 216-1019 216-1020 216-1012 216-1014 216-1016 216-1018
Sample containers, straight, with screw cap, WHEATON®
Sample Containers General Purpose Containers
Polystyrene clear, with metal screw cap.
  • Manufactured from FDA approved virgin polystyrene
  • Supplied sterile and for in vitro use only
  • CE marked in accordance with European Directive 98/79/EC
  • All containers are leak tested to British Standard BS5213:1975(1989)
  • All materials are non cytotoxic
Straight walled vessels ideal for sample collection, storage and transport. Polystyrene offers good thermal and chemical resistance. Supplied with flow seal metal cap.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904